ATC311768191 Atlantic Research Chemicals Ltd

1-[6-(3-Chlorophenyl)pyridin-3-yl]ethanone

  • MDL number MFCD12404913
  • Cas Number : 1216636-84-8
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002228
Availability details for
Quantity
  • Availability
  • Add to Cart