ATC311768213 Atlantic Research Chemicals Ltd

6-Amino-5-iodonicotinonitrile

  • MDL number MFCD12827898
  • Cas Number : 1187322-51-5
  • Molecular Weight : 245.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002248
Availability details for
Quantity
  • Availability
  • Add to Cart