ATC311768226 Atlantic Research Chemicals Ltd

2,4-Dimethyl-3-nitropyridine

  • MDL number MFCD08235269
  • Cas Number : 1074-76-6
  • Molecular Weight : 152.153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS00226
Availability details for
Quantity
  • Availability
  • Add to Cart