ATC311768318 Atlantic Research Chemicals Ltd

2-(5-Ethylpyridin-2-yl)ethanol

  • MDL number MFCD00129040
  • Cas Number : 5223-06-3
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00236
Availability details for
Quantity
  • Availability
  • Add to Cart