ATC311768342 Atlantic Research Chemicals Ltd

2-Methyl-3-nitropyridine hydrochloride

  • MDL number MFCD04114123
  • Cas Number : 63585-69-3
  • Molecular Weight : 174.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00262
Availability details for
Quantity
  • Availability
  • Add to Cart