ATC311768441 Atlantic Research Chemicals Ltd

Ammonium 2-Furyl(methoxyimino)acetate

  • MDL number MFCD03791040
  • Cas Number : 97148-39-5
  • Molecular Weight : 186.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00424
Availability details for
Quantity
  • Availability
  • Add to Cart