ATC311768477 Atlantic Research Chemicals Ltd

5-Methylisoxazole

  • MDL number MFCD00003152
  • Cas Number : 5765-44-6
  • Molecular Weight : 83.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00702
Availability details for
Quantity
  • Availability
  • Add to Cart