ATC311768501 Atlantic Research Chemicals Ltd

4-(1H-Pyrazol-4-yl)but-3-yn-1-ol

  • MDL number MFCD09800602
  • Cas Number : 175474-15-4
  • Molecular Weight : 136.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00909
Availability details for
Quantity
  • Availability
  • Add to Cart