ATC311768503 Atlantic Research Chemicals Ltd

1-Ethyl-1H-pyrazole

  • MDL number MFCD03444367
  • Cas Number : 2817-71-2
  • Molecular Weight : 96.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS00911
Availability details for
Quantity
  • Availability
  • Add to Cart