ATC311768562 Atlantic Research Chemicals Ltd

1H-Imidazole-4-carboxamide

  • MDL number MFCD04113065
  • Cas Number : 26832-08-6
  • Molecular Weight : 111.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS01014
Availability details for
Quantity
  • Availability
  • Add to Cart