ATC311768573 Atlantic Research Chemicals Ltd

2,4-Diphenyl-1H-imidazole

  • MDL number MFCD07772899
  • Cas Number : 670-83-7
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS01025
Availability details for
Quantity
  • Availability
  • Add to Cart