ATC311768607 Atlantic Research Chemicals Ltd

Pyrimidine-2-carbohydrazide

  • MDL number MFCD00508242
  • Cas Number : 87362-28-5
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS01207
Availability details for
Quantity
  • Availability
  • Add to Cart