ATC311768655 Atlantic Research Chemicals Ltd

3-Hydrazino-6-methylpyridazine

  • MDL number MFCD00067771
  • Cas Number : 38956-79-5
  • Molecular Weight : 124.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS01312
Availability details for
Quantity
  • Availability
  • Add to Cart