ATC311768758 Atlantic Research Chemicals Ltd

5-Chloro-1,3-dihydro-2H-benzimidazol-2-one

  • MDL number MFCD00493866
  • Cas Number : 2034-23-3
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS02209
Availability details for
Quantity
  • Availability
  • Add to Cart