ATC311768761 Atlantic Research Chemicals Ltd

6-Fluoro-1,3-dihydro-2H-benzimidazol-2-one

  • MDL number MFCD05663528
  • Cas Number : 1544-75-8
  • Molecular Weight : 152.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS02212
Availability details for
Quantity
  • Availability
  • Add to Cart