ATC311768773 Atlantic Research Chemicals Ltd

Decahydroquinoline

  • MDL number MFCD00006695
  • Cas Number : 2051-28-7
  • Molecular Weight : 139.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS02608
Availability details for
Quantity
  • Availability
  • Add to Cart