ATC311768781 Atlantic Research Chemicals Ltd

(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD08692036
  • Cas Number : 118864-75-8
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS02706
Availability details for
Quantity
  • Availability
  • Add to Cart