ATC311768797 Atlantic Research Chemicals Ltd

4-[(3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid

  • MDL number MFCD00269588
  • Cas Number : 857492-98-9
  • Molecular Weight : 283.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS02907
Availability details for
Quantity
  • Availability
  • Add to Cart