ATC311768852 Atlantic Research Chemicals Ltd

(4R,5S)-4-Methyl-5-phenyl-3-piperidin-4-yl-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD09991844
  • Cas Number : 1217459-01-2
  • Molecular Weight : 296.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS03743
Availability details for
Quantity
  • Availability
  • Add to Cart