ATC311768860 Atlantic Research Chemicals Ltd

tert-Butyl 4-(Ethylamino)piperidine-1-carboxylate

  • MDL number MFCD06808580
  • Cas Number : 264905-39-7
  • Molecular Weight : 228.334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS03751
Availability details for
Quantity
  • Availability
  • Add to Cart