ATC311768867 Atlantic Research Chemicals Ltd

5-Chloro-1-piperidin-4-yl-1,3-dihydro-2H-benzimidazol-2-one

  • MDL number MFCD02093793
  • Cas Number : 53786-28-0
  • Molecular Weight : 251.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS03758
Availability details for
Quantity
  • Availability
  • Add to Cart