ATC311768879 Atlantic Research Chemicals Ltd

3-Methoxypiperidin-2-one

  • MDL number MFCD12026381
  • Cas Number : 25219-59-4
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS03770
Availability details for
Quantity
  • Availability
  • Add to Cart