ATC311768931 Atlantic Research Chemicals Ltd

1-(5-Chloro-2-methylphenyl)piperazine

  • MDL number MFCD00087128
  • Cas Number : 76835-20-6
  • Molecular Weight : 210.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS03832
Availability details for
Quantity
  • Availability
  • Add to Cart