ATC311768946 Atlantic Research Chemicals Ltd

1-(Cyclopropylcarbonyl)piperazine hydrochloride

  • MDL number MFCD03428576
  • Cas Number : 59878-57-8
  • Molecular Weight : 190.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS03847
Availability details for
Quantity
  • Availability
  • Add to Cart