ATC311769049 Atlantic Research Chemicals Ltd

Cis-Cyclobutane-1,2-dicarbonitrile

  • MDL number MFCD00238568
  • Cas Number : 3211-19-6
  • Molecular Weight : 106.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS099111
Availability details for
Quantity
  • Availability
  • Add to Cart