ATC311769068 Atlantic Research Chemicals Ltd

1-Pyridin-2-ylcyclopropanecarbonitrile

  • MDL number MFCD15526928
  • Cas Number : 162960-28-3
  • Molecular Weight : 144.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS002293
Availability details for
Quantity
  • Availability
  • Add to Cart