ATC311769149 Atlantic Research Chemicals Ltd

2,2-Dimethylcyclopropanecarboxamide

  • MDL number MFCD00829406
  • Cas Number : 1759-55-3
  • Molecular Weight : 113.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS09978
Availability details for
Quantity
  • Availability
  • Add to Cart