ATC311769160 Atlantic Research Chemicals Ltd

4-Methyl-1,4-diazepan-5-one

  • MDL number MFCD09054767
  • Cas Number : 172314-56-6
  • Molecular Weight : 128.174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS09989
Availability details for
Quantity
  • Availability
  • Add to Cart