ATC311769204 Atlantic Research Chemicals Ltd

1-Naphthylacetonitrile

  • MDL number MFCD00004041
  • Cas Number : 132-75-2
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS100124
Availability details for
Quantity
  • Availability
  • Add to Cart