ATC311769209 Atlantic Research Chemicals Ltd

(1R,5S)-8-Benzyl-8-azabicyclo[3.2.1]octan-3-one

  • MDL number MFCD00151640
  • Cas Number : 28957-72-4
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS100129
Availability details for
Quantity
  • Availability
  • Add to Cart