ATC311769222 Atlantic Research Chemicals Ltd

1,5,6,7-Tetrahydro-4H-cyclopenta[d]pyrimidin-4-one

  • MDL number MFCD11977237
  • Cas Number : 5661-01-8
  • Molecular Weight : 136.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS100140
Availability details for
Quantity
  • Availability
  • Add to Cart