ATC311769279 Atlantic Research Chemicals Ltd

7H-Pyrrolo[2,3-d]pyrimidine-2,4-diol

  • MDL number MFCD03092515
  • Cas Number : 39929-79-8
  • Molecular Weight : 151.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS10023
Availability details for
Quantity
  • Availability
  • Add to Cart