ATC311769307 Atlantic Research Chemicals Ltd

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride

  • MDL number MFCD17926456
  • Cas Number : 54745-74-3
  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS10052
Availability details for
Quantity
  • Availability
  • Add to Cart