ATC311769368 Atlantic Research Chemicals Ltd

tert-Butyl 2-Aminoethyl(ethyl)carbamate hydrochloride

  • MDL number MFCD06796900
  • Cas Number : 1001020-32-1
  • Molecular Weight : 224.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS101104
Availability details for
Quantity
  • Availability
  • Add to Cart