ATC311769377 Atlantic Research Chemicals Ltd

2-Hydroxyacetohydrazide

  • MDL number MFCD09840691
  • Cas Number : 3530-14-1
  • Molecular Weight : 90.081

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=XS101112
Availability details for
Quantity
  • Availability
  • Add to Cart