ATC311769428 Atlantic Research Chemicals Ltd

Ethyl Imidocarbamate hydrochloride

  • MDL number MFCD07438010
  • Cas Number : 31407-74-6
  • Molecular Weight : 124.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS10146
Availability details for
Quantity
  • Availability
  • Add to Cart