ATC311769438 Atlantic Research Chemicals Ltd

Sodium N,N-Diethyldithiocarbamate (25% solution in water)

  • MDL number MFCD00041028
  • Cas Number : 148-18-5
  • Molecular Weight : 171.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS10157
Availability details for
Quantity
  • Availability
  • Add to Cart