ATC311769465 Atlantic Research Chemicals Ltd

4-(Dimethylamino)-1,1-dimethoxybut-3-en-2-one

  • MDL number MFCD00085024
  • Cas Number : 67751-23-9
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS10185
Availability details for
Quantity
  • Availability
  • Add to Cart