ATC311769473 Atlantic Research Chemicals Ltd

Methyl 4-Chlorobutanimidoate hydrochloride

  • MDL number MFCD12404920
  • Cas Number : 77570-15-1
  • Molecular Weight : 172.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=XS10193
Availability details for
Quantity
  • Availability
  • Add to Cart