ATC359509889 Atlantic Research Chemicals Ltd

5,6-Diphenyl-1,2,4-triazin-3-amine

  • MDL number MFCD00047461
  • Cas Number : 4511-99-3
  • Molecular Weight : 248.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE05302
Availability details for
Quantity
  • Availability
  • Add to Cart