ATC359509930 Atlantic Research Chemicals Ltd

2-Chloro-2',5'-difluoroacetophenone

  • MDL number MFCD04115827
  • Cas Number : 60468-36-2
  • Molecular Weight : 190.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=AB00116
Availability details for
Quantity
  • Availability
  • Add to Cart