ATC359509957 Atlantic Research Chemicals Ltd

1-(1,1'-Biphenyl-4-yl)-2-chloroethanone

  • MDL number MFCD00058937
  • Cas Number : 635-84-7
  • Molecular Weight : 230.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AB09902
Availability details for
Quantity
  • Availability
  • Add to Cart