ATC359510040 Atlantic Research Chemicals Ltd

(2R)-2-[(tert-Butoxycarbonyl)amino]-3-(1,3-thiazol-4-yl)propanoic acid

  • MDL number MFCD00079680
  • Cas Number : 134107-69-0
  • Molecular Weight : 272.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA00697
Availability details for
Quantity
  • Availability
  • Add to Cart