ATC359510041 Atlantic Research Chemicals Ltd

(2-Amino-1,3-thiazol-4-yl)[(2-tert-butoxy-2-oxoethoxy)imino]acetic acid

  • MDL number MFCD00071569
  • Cas Number : 86299-47-0
  • Molecular Weight : 301.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA00698
Availability details for
Quantity
  • Availability
  • Add to Cart