ATC359510052 Atlantic Research Chemicals Ltd

2-(Acetylamino)-3-(1H-imidazol-4-yl)propanoic acid hydrate

  • MDL number MFCD00150969
  • Cas Number : 213178-97-3
  • Molecular Weight : 215.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA01019
Availability details for
Quantity
  • Availability
  • Add to Cart