ATC359510074 Atlantic Research Chemicals Ltd

1-(4-Methylbenzyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD01934736
  • Cas Number : 96449-91-1
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA04598
Availability details for
Quantity
  • Availability
  • Add to Cart