ATC359510082 Atlantic Research Chemicals Ltd

4-(Aminomethyl)cyclohexanecarboxylic acid

  • MDL number MFCD00064951
  • Cas Number : 1197-18-8
  • Molecular Weight : 157.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=CA09988
Availability details for
Quantity
  • Availability
  • Add to Cart