ATC359510089 Atlantic Research Chemicals Ltd

(1R,3S,4R)-2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]heptane-3-carboxylic acid

  • MDL number MFCD21333140
  • Cas Number : 1932212-21-9
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA09995
Availability details for
Quantity
  • Availability
  • Add to Cart