ATC359510145 Atlantic Research Chemicals Ltd

2-Chloro-6-fluoroaniline

  • MDL number MFCD00040309
  • Cas Number : 363-51-9
  • Molecular Weight : 145.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=AE001209
Availability details for
Quantity
  • Availability
  • Add to Cart