ATC359510172 Atlantic Research Chemicals Ltd

Ethyl 2,4-Dichloro-6-methylnicotinate

  • MDL number MFCD00173918
  • Cas Number : 86129-63-7
  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00298
Availability details for
Quantity
  • Availability
  • Add to Cart